n-Decyl acetate, 98%
n-Decyl acetate, 98%
n-Decyl acetate, 98%
Thermo Scientific Chemicals

n-Decyl acetate, 98%

CAS: 112-17-4 | C12H24O2 | 200.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A18814.30
also known as A18814-30
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Chemical Identifiers
CAS112-17-4
IUPAC Namedecyl acetate
Molecular FormulaC12H24O2
InChI KeyNUPSHWCALHZGOV-UHFFFAOYSA-N
SMILESCCCCCCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Refractive Index1.4250-1.4290 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only