1-Methylcyclopentene, 98%
1-Methylcyclopentene, 98%
1-Methylcyclopentene, 98%
1-Methylcyclopentene, 98%
Thermo Scientific Chemicals

1-Methylcyclopentene, 98%

CAS: 693-89-0 | C6H10 | 82.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18816.22
also known as A18816-22
Price (USD)
761.65
Special offer
Online exclusive
Ends: 30-Jun-2026
896.00
Save 134.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
761.65
Special offer
Online exclusive
Ends: 30-Jun-2026
896.00
Save 134.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS693-89-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4300-1.4340 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only