1,8-Naphthalimide, 98%
1,8-Naphthalimide, 98%
1,8-Naphthalimide, 98%
Thermo Scientific Chemicals

1,8-Naphthalimide, 98%

CAS: 81-83-4 | C12H7NO2 | 197.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A18856.09
also known as A18856-09
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.60
Save 5.95 (16%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.60
Save 5.95 (16%)
Each
Chemical Identifiers
CAS81-83-4
IUPAC Name3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular FormulaC12H7NO2
InChI KeyXJHABGPPCLHLLV-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=C3C(C=CC=C13)=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormPowder or crystalline powder
Assay (HPLC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Rebecca M. Duke, et. al. 1Colorimetric and fluorescent anion sensors: an overview of recent developments in the use of 1,8-naphthalimide-based chemosensors.Chem. Soc. Rev.2010,39(10), 3936-3953.
  2. B. Ramachandram, et. al. Unusually High Fluorescence Enhancement of Some 1,8-Naphthalimide Derivatives Induced by Transition Metal Salts.J. Phys. Chem. B,2000,104(49), 11824-11832.