3-Methylvaleric acid, 99%
3-Methylvaleric acid, 99%
3-Methylvaleric acid, 99%
Thermo Scientific Chemicals

3-Methylvaleric acid, 99%

CAS: 105-43-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18886.22
also known as A18886-22
Price (USD)
326.65
Online Exclusive
363.00
Save 36.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
326.65
Online Exclusive
363.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS105-43-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Refractive Index1.4140-1.4170 @ 20?C
Appearance (Color)Clear colorless
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only