6-Aminophthalide, 95%
6-Aminophthalide, 95%
6-Aminophthalide, 95%
Thermo Scientific Chemicals

6-Aminophthalide, 95%

CAS: 57319-65-0 | C8H7NO2 | 149.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2.5 g
10 g
Catalog number A18896.02
also known as A18896-02
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.00
Save 11.35 (14%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.00
Save 11.35 (14%)
Each
Chemical Identifiers
CAS57319-65-0
IUPAC Name6-amino-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H7NO2
InChI KeyZIJZDNKZJZUROE-UHFFFAOYSA-N
SMILESNC1=CC=C2COC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
FormPowder
Appearance (Color)Pale cream to cream to yellow to pale brown
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

Solubility
Soluble in water 1360 mg/L 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. ProQuest.Design, Synthesis and Polymerization of Highly Branched Pseudodendrimers Through Tandem Reactions and Kinetics Measurements of Pseudodendrimeric Polymerization,1; ProQuest; USA, 2008; 85-103.
  2. P. Sutter.; C.D. Weis. The synthesis of monosubstituted derivatives of 6-amino-3,3'-bis(p-dimethylaminophenyl)phthalide.Dyes and Pigments.1990,12(4), 287-300.