Bis(4-chlorophenyl) sulfone, 99%
Bis(4-chlorophenyl) sulfone, 99%
Bis(4-chlorophenyl) sulfone, 99%
Thermo Scientific Chemicals

Bis(4-chlorophenyl) sulfone, 99%

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A18938.22
also known as A18938-22
Price (USD)
33.60
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
33.60
Each
Chemical Identifiers
CAS80-07-9
IUPAC Name1-chloro-4-(4-chlorobenzenesulfonyl)benzene
Molecular FormulaC12H8Cl2O2S
InChI KeyGPAPPPVRLPGFEQ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)145.0-152.0?C
Identification (FTIR)Conforms
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Laskoski, M.; Keller, T. M.; Ricks-Laskoski, H. L.; Giller, C. B.; Hervey, J. Improved Synthesis of Oligomeric Sulfone-Based Phthalonitriles. Macromol. Chem. Phys. 2015, 216 (17), 1808-1815.
  2. Abbasi, F.; Mehdipour-Ataei, S.; Khademinejad, S. Novel type of highly soluble and thermally stable poly(sulfone ether imide)s. Des. Monomers Polym. 2015, 18 (8), 789-798.