1-Chloro-2-methyl-2-propanol, 98%
1-Chloro-2-methyl-2-propanol, 98%
1-Chloro-2-methyl-2-propanol, 98%
Thermo Scientific Chemicals

1-Chloro-2-methyl-2-propanol, 98%

CAS: 558-42-9 | C4H9ClO | 108.57 g/mol
Have Questions?
Catalog NumberQuantity
A18978.14
also known as A18978-14
25 g
Catalog number A18978.14
also known as A18978-14
Price (USD)
71.65
Online Exclusive
79.20
Save 7.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
71.65
Online Exclusive
79.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS558-42-9
IUPAC Name1-chloro-2-methylpropan-2-ol
Molecular FormulaC4H9ClO
InChI KeyJNOZGFXJZQXOSU-UHFFFAOYSA-N
SMILESCC(C)(O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4375-1.4415 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only