Isopropyl chloroacetate, 99%
Isopropyl chloroacetate, 99%
Isopropyl chloroacetate, 99%
Isopropyl chloroacetate, 99%
Thermo Scientific Chemicals

Isopropyl chloroacetate, 99%

CAS: 105-48-6 | C5H9ClO2 | 136.58 g/mol
Have Questions?
Catalog NumberQuantity
A19070.AC
also known as A19070-AC
25 mL
Catalog number A19070.AC
also known as A19070-AC
Price (USD)
30.20
Each
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)
30.20
Each
Add to cart
Chemical Identifiers
CAS105-48-6
IUPAC Namepropan-2-yl 2-chloroacetate
Molecular FormulaC5H9ClO2
InChI KeyVODRWDBLLGYRJT-UHFFFAOYSA-N
SMILESCC(C)OC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4180-1.4210 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only