What solvent should I use to reconstitute 7-Amino-4-Methylcoumarin, reference standard (Cat. No. A191)? Once reconstituted, can the product be aliquoted and stored at -20 degrees C without damaging the molecule?
The 7-Amino-4-Methylcoumarin, reference standard (Cat. No. A191) can be resuspended in DMF or DMSO at a concentration of at least 1 mM. The molecular weight is 175.19 g/mol.
Yes, the solution can be stored in aliquots at -20 degrees C for at least 6 months.
Find additional tips, troubleshooting help, and resources within our Protein Standards and Ladders Support Center.
Yes, the solution can be stored in aliquots at -20 degrees C for at least 6 months.
Find additional tips, troubleshooting help, and resources within our Protein Standards and Ladders Support Center.