N-Guanylurea sulfate, 97%
N-Guanylurea sulfate, 97%
N-Guanylurea sulfate, 97%
Thermo Scientific Chemicals

N-Guanylurea sulfate, 97%

CAS: 591-01-5 | C4H12N8O2·H2SO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A19106.18
also known as A19106-18
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS591-01-5
IUPAC Namebis((diaminomethylidene)urea); sulfuric acid
Molecular FormulaC4H14N8O6S
InChI KeyIATXFPUBPMZBPH-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC(N)=NC(N)=O.NC(N)=NC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (Dry wt. basis)
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤10.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only