2,5-Dibromoaniline, 97%
2,5-Dibromoaniline, 97%
2,5-Dibromoaniline, 97%
Thermo Scientific Chemicals

2,5-Dibromoaniline, 97%

CAS: 3638-73-1 | C6H5Br2N | 250.921 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19114.22
also known as A19114-22
Price (USD)
217.65
Online Exclusive
242.00
Save 24.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
217.65
Online Exclusive
242.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS3638-73-1
IUPAC Name2,5-dibromoaniline
Molecular FormulaC6H5Br2N
InChI KeyWRTAZRGRFBCKBU-UHFFFAOYSA-N
SMILESNC1=CC(Br)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to dark brown to black
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥97.5%
Melting Point (clear melt)51-57°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only