Anthrone, 95%
Anthrone, 95%
Anthrone, 95%
Thermo Scientific Chemicals

Anthrone, 95%

CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A19118.14
also known as A19118-14
Price (USD)
96.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
96.30
Each
Chemical Identifiers
CAS90-44-8
IUPAC Name9,10-dihydroanthracen-9-one
Molecular FormulaC14H10O
InChI KeyRJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or needles
Appearance (Color)Cream to yellow to pale brown
Assay (GC)≥94.0%
Identification (FTIR)Conforms
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Asamenew G.; Bisrat D.; Mazumder A.; Asres K. In vitro antimicrobial and antioxidant activities of anthrone and chromone from the latex of Aloe harlana Reynolds.Phytotherapy research.2011,25(12), 1756-1760.
  2. Malterud KE.; Farbrot TL.; Huse AE.; Sund RB. Antioxidant and radical scavenging effects of anthraquinones and anthrones.Pharmacology.1993,47(1), 77-85.