The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
M. -G. A. Shvekhgeimer. Synthesis of halogenopyridines (review). Chemistry of Heterocyclic Compounds. 1996, 32 (9), 987-1015.