3,5-Dihydroxy-4-methylbenzoic acid, 97%
3,5-Dihydroxy-4-methylbenzoic acid, 97%
3,5-Dihydroxy-4-methylbenzoic acid, 97%
Thermo Scientific Chemicals

3,5-Dihydroxy-4-methylbenzoic acid, 97%

CAS: 28026-96-2 | C8H7O4 | 167.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A19196.03
also known as A19196-03
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.50
Save 4.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.50
Save 4.85
Each
Chemical Identifiers
CAS28026-96-2
IUPAC Name3,5-dihydroxy-4-methylbenzoate
Molecular FormulaC8H7O4
InChI KeyKMRRXSZDSGYLCD-UHFFFAOYSA-M
SMILESCC1=C(O)C=C(C=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)> 96.0%
Appearance (Color)White to pale cream
Assay (Silylated GC)> 96.0%
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only