5-Bromothiophene-2-carboxylic acid, 98%
5-Bromothiophene-2-carboxylic acid, 98%
5-Bromothiophene-2-carboxylic acid, 98%
5-Bromothiophene-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5-Bromothiophene-2-carboxylic acid, 98%

CAS: 7311-63-9 | C5H2BrO2S | 206.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A19242.14
also known as A19242-14
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7311-63-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only