delta-Hexanolactone, 98%
delta-Hexanolactone, 98%
delta-Hexanolactone, 98%
Thermo Scientific Chemicals

delta-Hexanolactone, 98%

CAS: 823-22-3 | C6H10O2 | 114.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19274.14
also known as A19274-14
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.70
Save 8.05 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.70
Save 8.05 (14%)
Each
Chemical Identifiers
CAS823-22-3
IUPAC Name6-methyloxan-2-one
Molecular FormulaC6H10O2
InChI KeyRZTOWFMDBDPERY-UHFFFAOYNA-N
SMILESCC1CCCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.4505-1.4545 @ 20?C
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. MacLeod, A. J.; Pieris, N. M. Volatile flavor components of wood apple (Feronia limonia) and a processed product. J. Agric. Food Chem. 1981, 29 (1), 49-53.
  2. Henke, E.; Bornscheuer, U. Esterases from Bacillus subtilis and B. stearothermophilus share high sequence homology but differ substantially in their properties. Appl. Microbiol. Biotechnol. 2002, 60 (3), 320-326.