Dihydrojasmone, 97%
Dihydrojasmone, 97%
Dihydrojasmone, 97%
Thermo Scientific Chemicals

Dihydrojasmone, 97%

CAS: 1128-08-1 | C11H18O | 166.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19298.14
also known as A19298-14
Price (USD)
32.65
Online Exclusive
36.20
Save 3.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
36.20
Save 3.55 (10%)
Each
Chemical Identifiers
CAS1128-08-1
IUPAC Name3-methyl-2-pentylcyclopent-2-en-1-one
Molecular FormulaC11H18O
InChI KeyYCIXWYOBMVNGTB-UHFFFAOYSA-N
SMILESCCCCCC1=C(C)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4780-1.4820 @ 20?C
Assay (GC)≥96.0%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only