(3-Carboxypropyl)triphenylphosphonium bromide, 97%
(3-Carboxypropyl)triphenylphosphonium bromide, 97%
(3-Carboxypropyl)triphenylphosphonium bromide, 97%
(3-Carboxypropyl)triphenylphosphonium bromide, 97%
Thermo Scientific Chemicals

(3-Carboxypropyl)triphenylphosphonium bromide, 97%

CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A19302.14
also known as A19302-14
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.70
Save 10.05 (14%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.70
Save 10.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS17857-14-6
IUPAC Name(3-carboxypropyl)triphenylphosphanium bromide
Molecular FormulaC22H22BrO2P
InChI KeyNKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES[Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)243.0-252.0?C
FormPowder
Appearance (Color)White
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

Solubility
Soluble in water. soluble in ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. Yu Luo et. al. Synthesis and biological evaluation of piperamide analogues as HDAC inhibitors. Bioorganic & Medicinal Chemistry Letters,. 2011, 21 (16), 4844-4846 .
  2. Yong-Kang Zhang et. al. Synthesis and structure-activity relationships of novel benzoxaboroles as a new class of antimalarial agents. Bioorganic & Medicinal Chemistry Letters,. 2011, 21(2), 644-651 .