2-Bromomethyl-1,3-dioxolane, 97%
2-Bromomethyl-1,3-dioxolane, 97%
2-Bromomethyl-1,3-dioxolane, 97%
Thermo Scientific Chemicals

2-Bromomethyl-1,3-dioxolane, 97%

CAS: 4360-63-8 | C4H7BrO2 | 167.002 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A19312.14
also known as A19312-14
Price (USD)
66.65
Online Exclusive
73.40
Save 6.75 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
73.40
Save 6.75 (9%)
Each
Chemical Identifiers
CAS4360-63-8
IUPAC Name2-(bromomethyl)-1,3-dioxolane
Molecular FormulaC4H7BrO2
InChI KeyCKIIJIDEWWXQEA-UHFFFAOYSA-N
SMILESBrCC1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4800-1.4850 @ 20?C
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. McInroy, G. R.; Raiber, E. A.; Balasubramanian, S. Chemical biology of genomic DNA: minimizing PCR bias. Chem. Commun. 2014, 50, 12047-12049.
  2. Gao, F.; Deng, X. J.; Tang, Y.; Tang, J. P.; Yang, J.; Zhang, Y. M. A simple and efficient copper oxide-catalyzed Barbier-Grignard reaction of unactivated aryl or alkyl bromides with ester. Tetrahedron Lett. 2014, 55 (4), 880-883.