p-Toluenesulfonic acid sodium salt, 90+%
p-Toluenesulfonic acid sodium salt, 90+%
p-Toluenesulfonic acid sodium salt, 90+%
Thermo Scientific Chemicals

p-Toluenesulfonic acid sodium salt, 90+%

CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A19332.30
also known as A19332-30
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS657-84-1
IUPAC Namesodium 4-methylbenzene-1-sulfonate
Molecular FormulaC7H7NaO3S
InChI KeyKVCGISUBCHHTDD-UHFFFAOYSA-M
SMILES[Na+].CC1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤5.0% (U.S. sourced material)
Chloride content≤0.5%
Assay from Supplier's CofA≥90.0%
CommentMaterial sourced in the U.S. and in other countries
Water Content (Karl Fischer Titration)≤5.0% (non-U.S. sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only