Ethyl phenylacetate, 99%
Thermo Scientific Chemicals

Ethyl phenylacetate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A19356.22
also known as A19356-22
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.30
Save 4.65 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.30
Save 4.65 (14%)
Each
Chemical Identifiers
CAS101-97-3
IUPAC Nameethyl 2-phenylacetate
Molecular FormulaC10H12O2
InChI KeyDULCUDSUACXJJC-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4960-1.4990 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.