L-(+)-2-Phenylglycine, 98+%
L-(+)-2-Phenylglycine, 98+%
L-(+)-2-Phenylglycine, 98+%
L-(+)-2-Phenylglycine, 98+%
Thermo Scientific Chemicals

L-(+)-2-Phenylglycine, 98+%

CAS: 2935-35-5 | C8H9NO2 | 151.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A19360.22
also known as A19360-22
Price (USD)
80.65
Online Exclusive
89.30
Save 8.65 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.30
Save 8.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS2935-35-5
IUPAC Name2-amino-2-phenylacetic acid
Molecular FormulaC8H9NO2
InChI KeyZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILESNC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Optical Rotation157.0 ± 2.0? (C=1 in 1N HCl)
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only