Ethyl trans-4-decenoate, 97%
Ethyl trans-4-decenoate, 97%
Ethyl trans-4-decenoate, 97%
Thermo Scientific Chemicals

Ethyl trans-4-decenoate, 97%

CAS: 76649-16-6 | C12H22O2 | 198.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A19364.09
also known as A19364-09
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.50
Save 6.85 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.50
Save 6.85 (15%)
Each
Chemical Identifiers
CAS76649-16-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4355-1.4405 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only