4-Chloro-2-fluoro-1-iodobenzene, 98%
4-Chloro-2-fluoro-1-iodobenzene, 98%
4-Chloro-2-fluoro-1-iodobenzene, 98%
Thermo Scientific Chemicals

4-Chloro-2-fluoro-1-iodobenzene, 98%

CAS: 6797-79-1 | C6H3ClFI | 256.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19376.06
also known as A19376-06
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS6797-79-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale red to red to brown
Refractive Index1.6045-1.6085 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only