2-Benzoxazolinone, 98%
2-Benzoxazolinone, 98%
2-Benzoxazolinone, 98%
Thermo Scientific Chemicals

2-Benzoxazolinone, 98%

CAS: 59-49-4 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19378.22
also known as A19378-22
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS59-49-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Loss on Drying≤1% (105°C/constant weight)
Melting Point (clear melt)132.0-142.0°C
Appearance (Color)White to cream to pale brown
FormCrystals or crystalline powder or powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only