2,4,5-Trichlorobenzenesulfonyl chloride, 98%
2,4,5-Trichlorobenzenesulfonyl chloride, 98%
2,4,5-Trichlorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2,4,5-Trichlorobenzenesulfonyl chloride, 98%

CAS: 15945-07-0 | C6H2Cl4O2S | 279.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number A19390.06
also known as A19390-06
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
36.80
Save 5.15 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
36.80
Save 5.15 (14%)
Each
Chemical Identifiers
CAS15945-07-0
IUPAC Name2,4,5-trichlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H2Cl4O2S
InChI KeyWNVVRCKTQSCPAC-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(C=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)64.0-70.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only