5-Bromo-2-methoxybenzaldehyde, 98+%
5-Bromo-2-methoxybenzaldehyde, 98+%
5-Bromo-2-methoxybenzaldehyde, 98+%
Thermo Scientific Chemicals

5-Bromo-2-methoxybenzaldehyde, 98+%

CAS: 25016-01-7 | C8H7BrO2 | 215.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19396.14
also known as A19396-14
Price (USD)
21.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
21.60
Each
Chemical Identifiers
CAS25016-01-7
IUPAC Name5-bromo-2-methoxybenzaldehyde
Molecular FormulaC8H7BrO2
InChI KeyIJIBRSFAXRFPPN-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Free acid (titration)≤1.0%
Melting Point (clear melt)112.5-118.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only