4-tert-Butylphenoxyacetyl chloride, 98%
4-tert-Butylphenoxyacetyl chloride, 98%
4-tert-Butylphenoxyacetyl chloride, 98%
Thermo Scientific Chemicals

4-tert-Butylphenoxyacetyl chloride, 98%

CAS: 90734-55-7 | C12H15ClO2 | 226.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A19408.09
also known as A19408-09
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.90
Save 8.25 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.90
Save 8.25 (14%)
Each
Chemical Identifiers
CAS90734-55-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5145-1.5185 @ 20?C
FormLiquid
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.
RUO – Research Use Only
Taichi Arai,; Sakiho Kobayashi,; Natsuko Oshimi,; Tetsutaro Igarashi,; Tadamitsu Sakurai. Photoinduced Large Increase in the Refractive Index of N-2-Thenoyloxyaryl-4-tert-butylphenoxyacetamide Film. Bulletin of the Chemical Society of Japan. 2013, 86(9), 1079-1081.