Bromocresol Purple
Bromocresol Purple
Bromocresol Purple
Thermo Scientific Chemicals

Bromocresol Purple

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A19530.18
also known as A19530-18
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS115-40-2
IUPAC Name3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H16Br2O5S
InChI KeyABIUHPWEYMSGSR-UHFFFAOYSA-N
SMILESCC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to light purple to pink to peach to yellow-tan powder
Color ChangeGreen-yellow @ pH 5.2 to Bluish-violet @ pH 6.8
Loss on Drying≤1.5% (105°C/constant weight)
Solution Test1% in ethanol: clear, yellow to orange
Specific Absorbance (E 1%/1cm)≥380 @ pH 5.2 (431nm)
View more
It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

Solubility
Insoluble in water. Soluble in ethanol (80 mg/ml), alcohol, methanol (4 mg/ml), methyl cellosolve (300 mg/ml) and dilute alkalies.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)
RUO – Research Use Only

General References:

  1. Hill PG, et al. Bromocresol purple and the measurement of albumin. Falsely high plasma albumin concentrations eliminated by increased reagent ionic strength.Ann Clin Biochem.1983,20(5), 264-70.
  2. Ueno T.; Hirayama S.; Sugihara M.; Miida T. ANNALS EXPRESS: The bromocresol green assay, but not the modified bromocresol purple assay, overestimates the serum albumin concentration in nephrotic syndrome through reaction with α2-macroglobulin.Ann. Clin. Biochem.2015.