1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- P.-W. Wang. A solid-state transformation in 1,5-dichloroanthraquinone. J. Appl. Cryst. 1979, 12, 239
- Edward H. Ruediger.; Magdy L. Kaldas.; Sham S. Gandhi.; Cristi Fedryna.; Martin S. Gibson. Reactions of 1,5-dichloroanthraquinone with nucleophiles. J. Org. Chem. 1980, 45, (10), 1974-1978