It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.
Solubility
Soluble in water at 20°C 5.9g/L. Solubility in hot methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Benke BP.; Madhavan N. Active ion transporters from readily accessible acyclic octapeptides containing 3-aminobenzoic acid and alanine.Chem. Commun.2013,49(66), 7340-7342.
- Yamamoto A. Absorption and metabolic characteristics of p-aminobenzoic acid and its isomer, m-aminobenzoic acid, from the rat small intestineJ. Pharm. Sci.;1991,1067-1071.