3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%
3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%
3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%

CAS: 85068-32-2 | C10H5F6N | 253.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A19576.06
also known as A19576-06
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS85068-32-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4205-1.4255 @ 20?C
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Murat Altunkaya.; Ergun Gonca. Synthesis and characterization of new soluble fluorinated seco-porphyrazines. Polyhedron. 2011, 30, (6), 1035-1039
  2. Chuandong Dou.; Dong Chen.; Javed Iqbal.; Yang Yuan.; Hongyu Zhang.; Yue Wang. Multistimuli-Responsive Benzothiadiazole-Cored Phenylene Vinylene Derivative with Nanoassembly Properties. Langmuir. 2011, 27, (10), 6323-6329