N,N'-Diphenylthiourea, 98%
N,N'-Diphenylthiourea, 98%
N,N'-Diphenylthiourea, 98%
N,N'-Diphenylthiourea, 98%
Thermo Scientific Chemicals

N,N'-Diphenylthiourea, 98%

CAS: 102-08-9 | C13H12N2S | 228.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A19644.0E
also known as A19644-0E
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS102-08-9
IUPAC Name1,3-diphenylthiourea
Molecular FormulaC13H12N2S
InChI KeyFCSHMCFRCYZTRQ-UHFFFAOYSA-N
SMILESS=C(NC1=CC=CC=C1)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)147.0-154.0?C
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
N,N'-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. G. Yagupsky.; R. Levitus. N,N'-Dicyclohexylthiourea and N,N'-Diphenylthiourea Complexes of Cobalt(II). Inorg. Chem. 1965, 4, (11), 1589-1594 
  2. N. Yamazaki.; T. Iguchi.; F. Higashi. Studies on reactions of the N-phosphonium salts of pyridines—XV : Direct carbonylation of amines with carbon dioxide by a hydrolysis-dehydration reaction with phosphorus compounds. Tetrahedron. 1975, 31, (24), 3031-3034