2,4,6-Trihydroxybenzaldehyde, 95%
2,4,6-Trihydroxybenzaldehyde, 95%
2,4,6-Trihydroxybenzaldehyde, 95%
2,4,6-Trihydroxybenzaldehyde, 95%
Thermo Scientific Chemicals

2,4,6-Trihydroxybenzaldehyde, 95%

CAS: 487-70-7 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A19678.06
also known as A19678-06
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS487-70-7
IUPAC Name2,4,6-trihydroxybenzaldehyde
Molecular FormulaC7H6O4
InChI KeyBTQAJGSMXCDDAJ-UHFFFAOYSA-N
SMILESOC1=CC(O)=C(C=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown or pink
Identification (FTIR)Conforms
FormPowder
Assay (HPLC)≥94.0%

2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Ayers, S.; Shi, Z.; Marshall, J.; Fenster, M.; Huang, Y.; Pathirana, C. An unexpected product from the attempted acetal formation of 2,4,6-trihydroxybenzaldehyde. Tetrahedron Lett. 2015, 56 (36), 5132-5134.
  2. Suthar, S. K.; Aggarwal, V.; Chauhan, M.; Sharma, A.; Bansal, S.; Sharma, M. Molecular docking and biological evaluation of hydroxy-substituted (Z)-3-benzylideneindolin-2-one chalcones for the lead identification as tyrosinase inhibitors. Med. Chem. Res. 2015, 24 (3), 1331-1341.