Bis(methylthio)methane, 99%
Bis(methylthio)methane, 99%
Bis(methylthio)methane, 99%
Bis(methylthio)methane, 99%
Thermo Scientific Chemicals

Bis(methylthio)methane, 99%

CAS: 1618-26-4 | C3H8S2 | 108.217 g/mol
Have Questions?
Catalog NumberQuantity
A19706.14
also known as A19706-14
25 g
Catalog number A19706.14
also known as A19706-14
Price (USD)
81.65
Online Exclusive
90.70
Save 9.05 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
81.65
Online Exclusive
90.70
Save 9.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS1618-26-4
IUPAC Namebis(methylsulfanyl)methane
Molecular FormulaC3H8S2
InChI KeyLOCDPORVFVOGCR-UHFFFAOYSA-N
SMILESCSCSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5325-1.5355 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only