2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%
2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%
2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%
2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%

CAS: 28286-86-4 | C7H5Cl3O2S | 259.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Catalog number A19718.18
also known as A19718-18
Price (USD)
675.65
Online Exclusive
750.00
Save 74.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
675.65
Online Exclusive
750.00
Save 74.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS28286-86-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to cream
Melting Point (clear melt)67.0-76.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only