gamma-Decanolactone, 98%
gamma-Decanolactone, 98%
gamma-Decanolactone, 98%
Thermo Scientific Chemicals

gamma-Decanolactone, 98%

CAS: 706-14-9 | C10H18O2 | 170.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19732.14
also known as A19732-14
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.50
Save 10.85 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.50
Save 10.85 (14%)
Each
Chemical Identifiers
CAS706-14-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4470-1.4510 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only