2'-Fluoro-4'-hydroxyacetophenone, 97%
2'-Fluoro-4'-hydroxyacetophenone, 97%
2'-Fluoro-4'-hydroxyacetophenone, 97%
Thermo Scientific Chemicals

2'-Fluoro-4'-hydroxyacetophenone, 97%

CAS: 98619-07-9 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19884.06
also known as A19884-06
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS98619-07-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Pale brown or grey
FormPowder
Melting Point (clear melt)117.0-126.0?C
2'-Fluoro-4'-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-4′-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

General References:

  1. Xiao-Bo Zhang; Qun Lu & Wei-Ming Xu. A Practical Synthesis of 2'-Fluoro- 4'-hydroxyacetophenone. Organic Preparations and Procedures International: The New Journal for Organic Synthesis. 2009, 41, 419-421.