Brucine sulfate hydrate, 98% (dry wt.), water <13%
Brucine sulfate hydrate, 98% (dry wt.), water <13%
Brucine sulfate hydrate, 98% (dry wt.), water <13%
Thermo Scientific Chemicals

Brucine sulfate hydrate, 98% (dry wt.), water <13%

CAS: 652154-10-4 | C46H54N4O12S | 887.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19943.22
also known as A19943-22
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS652154-10-4
IUPAC Namebis((1R,11S,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one); sulfuric acid
Molecular FormulaC46H54N4O12S
InChI KeyHCMSIGALSOEZRW-MDQLRNFDSA-N
SMILESOS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (dry wt. basis)
Water Content (Karl Fischer Titration)≤13%
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
A. M. Figueiredo Neto; M. E. Marcondes. Helene. Phase diagram of the cholesteric lyotropic mesophase potassium laurate-n-decanol-water-d2-brucine sulfate ferrofluid. J. Phys. Chem. 1987, 91, (6), 1466-1469.