Alexa Fluor™ 555 Protein Labeling Kit, 1 kit - FAQs
Alexa Fluor™ 555 Protein Labeling Kit, 1 kit - FAQs
View additional product information for Antibody Labeling Kits for 1 mg - FAQs (A10235, A10236, A10237, A10238, A20170, A10239, A20173, P30012, A20174, A20171, A20172, D20655, F10240, O10241, A10170)
3 product FAQs found
Can I use 50 μg of protein with Fluorescent Protein Labeling Kits?
No. We recommend using 1 mg of protein with Fluorescent Protein Labeling Kits. For smaller protein sample sizes, we recommend using Microscale Protein Labeling kits which are optimized for 20-100 µg of protein.
Find additional tips, troubleshooting help, and resources within our
Cell Analysis Support Center.
What formulation of antibody should I use for conjugation for small animal in vivo imaging?
To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.
Find additional tips, troubleshooting help, and resources within our
Cell Analysis Support Center.
What is degree of labeling (DOL)?
Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.
Find additional tips, troubleshooting help, and resources within our
Cell Analysis Support Center.