Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488 - FAQs
Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488 - FAQs
View additional product information for Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488 - FAQs (A21206)
2 product FAQs found
My Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor 488 (Cat. No. A21206) was shipped at ambient temperature. Is it still viable?
Yes, this product is supposed to be shipped at ambient temperature.
My Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor 555 (Cat. No. A21424) was shipped at ambient temperature. Is it still viable?
Yes, this product is supposed to be shipped at ambient temperature.