Amplex™ Red and Amplex™ UltraRed Reagents
Invitrogen™

Amplex™ Red and Amplex™ UltraRed Reagents

The Amplex Red and UltraRed reagents are fluorogenic substrates for peroxidase. These highly sensitive and stable probes react with H2O2 in the presence of HRP for solution-based assays and ELISAs. Amplex UltraRed is a brighter version of Amplex Red with an improved pH range and sensitivity.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityDye Type
A2217710 x 10 mgAmplex Red
A122225 mgAmplex Red
A360065 x 1 mgAmplex UltraRed
Catalog number A22177
Price (USD)
3,099.65
Online exclusive
3,195.00
Save 95.35
Each
Quantity:
10 x 10 mg
Dye Type:
Amplex Red
Price (USD)
3,099.65
Online exclusive
3,195.00
Save 95.35
Each
The Amplex Red and Amplex UltraRed reagents are highly sensitive and stable fluorescent probes for the detection of hydrogen peroxide (H2O2) and ideal fluorogenic substrates for peroxidase. These reagents can be used for solution-based assays to detect the activity of H2O2 released from biological samples or with HRP-based ELISA applications. The Amplex UltraRed reagent is an improved version of Amplex Red that is brighter and more sensitive to peroxidase with less pH sensitivity compared to the Amplex Red reagent.

In the presence of horseradish peroxidase (HRP), the Amplex Red and UltraRed reagents react in a 1:1 stoichiometry with H2O2 to produce highly fluorescent, strongly absorbing resorufin (ex/em ∼570/585 nm) or Amplex UltraRed (ex/em ∼568/581 nm), respectively. These oxidized reaction products have a broad red emission with little interference from blue and green autofluorescence found in most biological samples. Because H2O2 is produced in many different enzymatic reactions, the Amplex Red and UltraRed reagents coupled with HRP can be used to detect the activity of many different enzymes. These reagents are also used to detect H2O2 released from biological samples, including cells and isolated mitochondria.

Features of the Amplex Red reagent:

  • Sensitive and fast detection of HRP activity or H2O2 for solution-based assays and ELISAs
  • Detection of H2O2 produced in many different enzymatic reactions
  • Measurement with either fluorescence or absorbance, permitting either fluorogenic or chromogenic detection

The Amplex Red reagent has been used to develop a variety of novel fluorogenic and chromogenic assays for enzymes that produce hydrogen peroxide. When coupled with added horseradish peroxidase, these assays that generate H2O2 permit the ultrasensitive quantitation of a diverse assortment of analytes, including glucose, galactose, cholesterol, glutamic acid, xanthine (or hypoxanthine), uric acid, choline, and acetylcholine.

The Amplex UltraRed reagent offers brighter fluorescence and enhanced sensitivity on a per-mole basis in peroxidase or peroxidase-coupled enzyme assays compared to Amplex Red. The Amplex UltraRed reagent is also less sensitive to acidic pH and exhibits improved stability in the presence of H2O2 or thiols such as dithiothreitol (DTT). For HRP-based ELISA applications, this results in at least two-fold greater sensitivity compared to Amplex Red.

Benefits of the Amplex UltraRed reagent over Amplex Red:

  • Brighter signals and enhanced sensitivity for HRP activity and H2O2
  • Less sensitive to acidic pH
  • Improved stability in the presence of H2O2 or thiols such as DTT

The Amplex Red and Amplex UltraRed reaction products resorufin and Amplex UltroxRed have high extinction coefficients so they can be measured either fluorometrically or spectrophotometrically with absorbance at ∼560 nm, providing flexibility in detection of H2O2, various enzyme assays, and HRP-based ELISAs.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAmplex Red
Emission Wavelength585 nm
Excitation Wavelength571 nm
FormSolid
Quantity10 x 10 mg
Shipping ConditionRoom Temperature
SubstrateHRP (Horseradish Peroxidase) Substrate
Substrate TypeChemical substrate
Validated ApplicationCell Analysis
For Use With (Application)Enzyme and Metabolic Activity
For Use With (Equipment)Fluorometer, Microplate Reader
Product LineAmplex
Product TypeRed Hydrogen Peroxide
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).

Frequently asked questions (FAQs)

I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I have ordered and tested your Amplex Red Reagent (Cat. Nos. A12222, A22177), but it is not reacting correctly. What could be the cause?

Either the horseradish peroxidase or other enzymes in the assay may not be active, the H2O2 may have degraded, or the sample could have some component that is consuming H2O2 or inactivating the enzymes required for the assay. We recommend that you check when the reagent was purchased and how it was stored prior to use; the reagent should not be used past the warranty period.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you recommend using Amplex Red Reagent (Cat. Nos. A12222, A22177) in live cells?

Amplex Red is cell impermeant, but also its conversion to resorufin requires direct contact with the reaction of peroxidase with H2O2 (in solution, at the surface, or within cells). Within cells, peroxidases and H2O2 may not be sufficiently localized or active to provide enough signal.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use Amplex Red Reagent (Cat. Nos. A12222, A22177) in the presence of catalases?

Unless you are specifically analyzing catalase activity, as with the Amplex Red Catalase Assay Kit (Cat. No. A22180), the assay should be devoid of catalases. Catalases compete with the horseradish peroxidase for H2O2; the conversion of Amplex Red to resorufin requires the reaction of horseradish peroxidase with H2O2.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How should I store Amplex Red Reagent (Cat. Nos. A12222, A22177)?

We recommend storing Amplex Red Reagent under vacuum (anaerobic conditions) and dehydrated. Do not store Amplex Red Reagent in solution.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (224)

Citations & References
Abstract
Roles for the human ATP-dependent Lon protease in mitochondrial DNA maintenance.
Authors:Lu B,Yadav S,Shah PG,Liu T,Tian B,Pukszta S,Villaluna N,Kutejová E,Newlon CS,Santos JH,Suzuki CK
Journal:The Journal of biological chemistry
PubMed ID:17420247
Pressure overload-induced mild cardiac hypertrophy reduces left ventricular transmural differences in mitochondrial respiratory chain activity and increases oxidative stress.
Authors:Kindo M, Gerelli S, Bouitbir J, Charles AL, Zoll J, Hoang Minh T, Monassier L, Favret F, Piquard F, Geny B,
Journal:Front Physiol
PubMed ID:22934079
Objective: Increased mechanical stress and contractility characterizes normal left ventricular (LV) subendocardium (Endo) but whether Endo mitochondrial respiratory chain complex activities is reduced as compared to subepicardium (Epi) and whether pressure overload-induced LV hypertrophy (LVH) might modulate transmural gradients through increased reactive oxygen species (ROS) production is unknown. Methods: LVH ... More
Overexpression of nicastrin increases Abeta production.
Authors:Murphy MP, Das P, Nyborg AC, Rochette MJ, Dodson MW, Loosbrock NM, Souder TM, McLendon C, Merit SL, Piper SC, Jansen KR, Golde TE
Journal:FASEB J
PubMed ID:12692078
Gamma-secretase cleavage is the final proteolytic step that releases the amyloid beta-peptide (Abeta) from the amyloid beta-protein precursor (APP). Significant evidence indicates that the presenilins (PS) are catalytic components of a high molecular weight gamma-secretase complex. The glycoprotein nicastrin was recently identified as a functional unit of this complex based ... More
Association of excitatory amino acid transporters, especially EAAT2, with cholesterol-rich lipid raft microdomains: importance for excitatory amino acid transporter localization and function.
Authors:Butchbach ME, Tian G, Guo H, Lin CL
Journal:J Biol Chem
PubMed ID:15187084
In the present study, we investigated the role of membrane cholesterol in the function of glutamate transporters. Depletion of membrane cholesterol by methyl-beta-cyclodextrin resulted in reduced Na(+)-dependent glutamate uptake in primary cortical cultures. Glial glutamate transporter EAAT2-mediated uptake was more sensitive to this effect. Cell surface biotinylation and immunostaining experiments ... More
The characterization of mitochondrial permeability transition in clonal pancreatic beta-cells. Multiple modes and regulation.
Authors:Koshkin V, Bikopoulos G, Chan CB, Wheeler MB
Journal:J Biol Chem
PubMed ID:15231823
Mitochondrial permeability transition (MPT), which contributes substantially to the regulation of normal mitochondrial metabolism, also plays a crucial role in the initiation of cell death. It is known that MPT is regulated in a tissue-specific manner. The importance of MPT in the pancreatic beta-cell is heightened by the fact that ... More