ATTO-TAG™ CBQCA Amine-Derivatization Kit - FAQs
ATTO-TAG™ CBQCA Amine-Derivatization Kit - FAQs
View additional product information for ATTO-TAG™ CBQCA Amine-Derivatization Kit - FAQs (A2333)
2 product FAQs found
Can CBQCA (3-(4carboxybenzoyl) quinoline- 2-carboxaldehyde) be used to label reducing sugars?
Yes, see the reference: Novotny MV, Sudor J (1993) High-performance capillary electrophoresis of glycoconjugates. Electrophoresis 14:373-389.
Can NaCN be used instead of KCN in the CBQCA (3-(4carboxybenzoyl) quinoline- 2-carboxaldehyde) assay?
Yes. Use the same molarity; it is the cyanide group that is required for the reaction.