Can I purchase the primary antibodies from the Human Neural Stem Cell Immunocytochemistry Kit (Cat. No. A24354) separately?
Yes, the primary antibodies from the Human Neural Stem Cell Immunocytochemistry Kit (Cat. No. A24354) can be purchased separately. Here are the catalog numbers:
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center
- Nestin Monoclonal Antibody (10C2), eBioscience (Cat. No. 14984382)
- SOX2 Polyclonal Antibody (Cat. No. 481400)
- PAX6 Polyclonal Antibody (Cat. No. 426600)
- SOX1 Goat anti-Human, Mouse, Rat, Polyclonal, R&D Systems; available through Fisher Scientific (Cat. No. AF3369)
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center