Can you provide references for growing adult neurons?
Instructions for growing adult neurons can be found in the Neurobasal-A Medium product insert. You can also refer to the following publication for growing adult neurons:
• Isolation and Culture of Adult Rat Hippocampal Neurons by Brewer, G.J., J. Neuroscience Methods 71 (1997), 145-158.
• Isolation and Culture of Adult Rat Hippocampal Neurons by Brewer, G.J., J. Neuroscience Methods 71 (1997), 145-158.