Attune™ Wash Solution, 250 mL - FAQs
Attune™ Wash Solution, 250 mL - FAQs
View additional product information for Attune™ Wash Solution - FAQs (J24974, A24974)
1 product FAQs found
I've run out of Attune Wash Solution for the Attune NxT Flow Cytometer. What can I use for the time being in place of the wash solution until I can get more wash solution?
In an emergency, we recommend using the highest purity of water available in place of the wash solution and then switching to the Attune Wash Solution as soon as possible.