Attune™ Shutdown Solution (1X), 250 mL - FAQs
Attune™ Shutdown Solution (1X), 250 mL - FAQs
View additional product information for Attune™ Shutdown Solution (1X) - FAQs (A24975, J106628)
1 product FAQs found
I've run out of Attune Shutdown Solution for the Attune NxT Flow Cytometer. What can I use for the time being in place of the shutdown solution until I can get more shutdowns solution?
In an emergency, we recommend using the highest purity of water available in place of the shutdown solution and then switching to the Attune Shutdown Solution as soon as possible.