Soy Peptone Halal, ultra-filtered (UF)
Gibco™
Soy Peptone Halal, ultra-filtered (UF)
Gibco Soy Peptone Halal UF is a halal-certified, animal-origin-free complex nutrient for vaccine applications, enabling high-yield, scalable growth with reliable lot-to-lot consistency and simplified halal compliance.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A2513802600 g
A251380110 kg
Número de catálogo A2513802
Precio (USD)
-
Cantidad:
600 g
Gibco Soy Peptone Halal UF is a halal-certified, animal-origin-free (AOF) soy peptone engineered for high-yield bacterial growth that helps accelerate vaccine and biologics development. It drives process consistency from early development through commercial scale-up while supporting robust cell health. AOF raw materials and IFANCA halal certification help de-risk raw materials, streamline compliance, and expand access to halal-conscious markets. Gibco Soy Peptone Halal UF integrates easily into existing media and regulatory filings.
Features of Gibco Soy Peptone Halal UF include:
  • Halal-certified & AOF—supports global market access and reduces regulatory complexity
  • Proven performance—enables growth comparable to or exceeding standard soy peptones for vaccine development
  • Consistent results—supports predictable culture performance from development through scale-up
  • Quality-controlled sourcing—helps reduce contamination risk and strengthen regulatory documentation
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Tipo de célulaMammalian, Insect, Microbial
Certificaciones/conformidadIFANCA Halal Product Certificate
Para utilizar conPreparing and supplementing bacterial, insect, and mammalian cell culture media
Cantidad600 g
ClasificaciónAnimal Origin-free, Serum-free
FormularioPowder
Tipo de productoCell Culture Supplement
EsterilidadNon-sterile
Con aditivosNone
Sin aditivosNo Animal Derived Components
Unit SizeEach