Proteinase K for MagMAX™ DNA Multi-Sample Ultra Kit
Applied Biosystems™
Proteinase K for MagMAX™ DNA Multi-Sample Ultra Kit
This is the same proteinase K found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. Nos. A25597, A25598), available separatelyRead more
Have Questions?
Catalog NumberQuantity
A255614 mL
Catalog number A25561
Price (USD)
426.65
Online exclusive
474.00
Save 47.35
Each
Quantity:
4 mL
Price (USD)
426.65
Online exclusive
474.00
Save 47.35
Each
This is the same proteinase K found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. Nos. A25597, A25598), available separately in case more proteinase K is required for a particular application.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)DNA Extraction
Product LineMagMAX
Product TypeProteinase K
Quantity4 mL
FormatKit
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
What is the concentration of Proteinase K enzyme in Proteinase K for MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25561)?
The Proteinase K is provided at a concentration of 100 mg/mL.
What type of samples is the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) optimized for?
We have optimized protocols for extracting DNA from buccal swabs, whole blood, blood cards (Whatman FTA), oral rinse, saliva, and urine samples. In addition, the kit can be used to process vaginal microbiota samples (e.g., ThinPrep, Surepath) for gDNA extraction from yeast, bacteria (Gram positive/negative), viruses, and protozoa with the addition of additional enzymatic steps.
In the MagMAX DNA Multi-Sample Ultra Kit protocol, why do blood samples appear cloudy after adding the Multi-Sample DNA Lysis Buffer?
On rare occasions, the sample may turn cloudy after adding the Multi-Sample DNA Lysis Buffer. The sample will clear up when isopropanol is added. This will not have any impact on the yield or quality of the DNA.
Can heparin be used to collect blood samples for the MagMAX DNA Multi-Sample Ultra Kit?
Heparin is not recommended as an anti-coagulant since it can cause inhibition of PCR reactions. Instead, we strongly recommend collecting blood samples in EDTA or sodium citrate.
What is the best way to remove the buccal swabs from the MagMAX DNA Multi-Sample Ultra Kit lysate?
We recommend transferring the lysate to a new MagMAX Express-96 Deep Well Plate (Cat. No. 4388476). This option eliminates contamination risks and saves time. To transfer lysates, set a multi-channel micropipetor to ˜300 µL and transfer one row at a time. Each well should contain 200-250 µL after transfer.