DNA Binding Beads for MagMAX™ DNA Multi-Sample Ultra Kit
Applied Biosystems™
DNA Binding Beads for MagMAX™ DNA Multi-Sample Ultra Kit
These are the same DNA Binding Beads found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. Nos. A25597, A25598), availableRead more
Have Questions?
Catalog NumberQuantity
A255628 mL
Catalog number A25562
Price (USD)
499.65
Online exclusive
532.00
Save 32.35
Each
Quantity:
8 mL
Price (USD)
499.65
Online exclusive
532.00
Save 32.35
Each
These are the same DNA Binding Beads found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. Nos. A25597, A25598), available separately in case more DNA Binding Beads are required for a particular application.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeDNA Binding Bead
Quantity8 mL
Sample TypeBlood, Buccal Samples, Cells, Liquid Samples (e.g. Serum), Tissue
TargetGenomic DNA
FormatKit
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C). DO NOT FREEZE.
Frequently asked questions (FAQs)
What type of samples is the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) optimized for?
We have optimized protocols for extracting DNA from buccal swabs, whole blood, blood cards (Whatman FTA), oral rinse, saliva, and urine samples. In addition, the kit can be used to process vaginal microbiota samples (e.g., ThinPrep, Surepath) for gDNA extraction from yeast, bacteria (Gram positive/negative), viruses, and protozoa with the addition of additional enzymatic steps.
In the MagMAX DNA Multi-Sample Ultra Kit protocol, why do blood samples appear cloudy after adding the Multi-Sample DNA Lysis Buffer?
On rare occasions, the sample may turn cloudy after adding the Multi-Sample DNA Lysis Buffer. The sample will clear up when isopropanol is added. This will not have any impact on the yield or quality of the DNA.
Can heparin be used to collect blood samples for the MagMAX DNA Multi-Sample Ultra Kit?
Heparin is not recommended as an anti-coagulant since it can cause inhibition of PCR reactions. Instead, we strongly recommend collecting blood samples in EDTA or sodium citrate.
What is the best way to remove the buccal swabs from the MagMAX DNA Multi-Sample Ultra Kit lysate?
We recommend transferring the lysate to a new MagMAX Express-96 Deep Well Plate (Cat. No. 4388476). This option eliminates contamination risks and saves time. To transfer lysates, set a multi-channel micropipetor to ˜300 µL and transfer one row at a time. Each well should contain 200-250 µL after transfer.
Can buccal swabs that are to be used with the MagMAX DNA Multi-Sample Ultra Kit be shipped after sample collection?
Buccal swabs can be shipped at 25 degrees C or below. Exposure of the swabs to high temperatures (over 30 degrees C) during shipment has been associated with poor performance in the workflow. To prevent degradation, ensure that buccal swabs are completely dry before shipment. Place the swabs in a paper envelope for storage after collection - paper envelopes are permeable and allow the swabs to dry effectively.